Merck
  • Tuning intramolecular anharmonic vibrational coupling in 4-nitroaniline by solvent-solute interaction.
Tuning intramolecular anharmonic vibrational coupling in 4-nitroaniline by solvent-solute interaction.
The Journal of chemical physics (2007-12-25)
J Dreyer, V Kozich, W Werncke
ABSTRACT
Applying a combined experimental and theoretical approach we demonstrate that doublets of the nu(s)(NO(2)) band of 4-nitroaniline which have been observed in several environments originate from Fermi resonances. Changes of the line shapes typical for Fermi resonances are reported also for other isotopomers of 4-nitroaniline, however, for each of them in different solvents and solvent mixtures. Simulations of the infrared spectra based on the solvatochromic frequency shifts of the nu(s)(NO(2)) vibration determined experimentally together with calculated cubic couplings with overtones and combination bands account for the experimental findings.
Product Description
Supelco
4-Nitroaniline, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mL
In Stock
Details...
€ 36,60
100 mL
Please contact Customer Service for Availability
€ 63,70
500 mL
In Stock
Details...
€ 82,10
1 L
Please contact Customer Service for Availability
€ 114,00
1 gal
Please contact Customer Service for Availability
€ 347,00
5 gal
Please contact Customer Service for Availability
€ 1.390,00
Sigma-Aldrich
4-Nitroaniline, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mL
In Stock
Details...
€ 36,60
100 mL
Please contact Customer Service for Availability
€ 63,70
500 mL
In Stock
Details...
€ 82,10
1 L
Please contact Customer Service for Availability
€ 114,00
1 gal
Please contact Customer Service for Availability
€ 347,00
5 gal
Please contact Customer Service for Availability
€ 1.390,00