Merck
  • The cooperative electrochemical oxidation of chlorophenols in anode-cathode compartments.
The cooperative electrochemical oxidation of chlorophenols in anode-cathode compartments.
Journal of hazardous materials (2007-11-13)
Hui Wang, Jian Long Wang
ABSTRACT
By using a self-made carbon/polytetrafluoroethylene (C/PTFE) O2-fed as the cathode and Ti/IrO2/RuO2 as the anode, the degradation of three organic compounds (phenol, 4-chlorophenol, and 2,4-dichlorophenol) was investigated in the diaphragm (with terylene as diaphragm material) electrolysis device by electrochemical oxidation process. The result indicated that the concentration of hydrogen peroxide (H2O2) was 8.3 mg/L, and hydroxyl radical (HO) was determined in the cathodic compartment by electron spin resonance spectrum (ESR). The removal efficiency for organic compounds reached about 90% after 120 min, conforming to the sequence of phenol, 4-chlorophenol, and 2,4-dichlorophenol. And the dechlorination degree of 4-chlorophenol exceeded 90% after 80 min. For H2O2, HO existed in the catholyte and reduction dechlorination at the cathode, the mineralization of organics in the cathodic compartment was better than that in the anodic compartment. The degradation of organics was supposed to be cooperative oxidation by direct or indirect electrochemical oxidation at the anode and H2O2, HO produced by oxygen reduction at the cathode. High-performance liquid chromatography (HPLC) allowed identifying phenol as the dechlorination product of 4-chlorophenol in the cathodic compartment, and hydroquinone, 4-chlorocatechol, benzoquinone, maleic, fumaric, oxalic, and formic acids as the main oxidation intermediates in the cathodic and anodic compartments. A reaction scheme involving all these intermediates was proposed.
Sigma-Aldrich
Ruthenium(IV) oxide hydrate, powder
Sigma-Aldrich
Ruthenium(IV) oxide, 99.9% trace metals basis
Sigma-Aldrich
Ruthenium(IV) oxide hydrate, 99.9% trace metals basis