Merck
  • Characterization of pyramidal inversion boundaries in Sb2O3-doped ZnO by using electron back-scattered diffraction (EBSD).
Characterization of pyramidal inversion boundaries in Sb2O3-doped ZnO by using electron back-scattered diffraction (EBSD).
Acta crystallographica. Section A, Foundations of crystallography (2007-04-17)
Wook Jo, Chan Park, Doh-Yeon Kim
ABSTRACT
The composition planes of the inversion boundary induced by the addition of Sb2O3 to ZnO ceramics were analyzed crystallographically by the application of electron back-scattered diffraction (EBSD) analysis and stereographic projection techniques. The inversion boundary was determined to consist of three discrete composition planes, {0001}, {1011}, {1010}.
Sigma-Aldrich
Antimony(III) oxide, powder, 5 μm, ReagentPlus®, 99%
Sigma-Aldrich
Antimony(III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Antimony(III) oxide, 99.999% trace metals basis
Sigma-Aldrich
Antimony(III) oxide, nanopowder, <250 nm particle size (TEM), ≥99.9% trace metals basis