Merck
  • Allergic contact dermatitis from octisalate and cis-3-hexenyl salicylate.
Allergic contact dermatitis from octisalate and cis-3-hexenyl salicylate.
Dermatitis : contact, atopic, occupational, drug (2006-09-08)
Daniel W Shaw
ABSTRACT
A 62-year-old woman developed allergic contact dermatitis from sunscreens containing octisalate (octyl salicylate, 2-ethylhexyl salicylate) and from a fragrance containing cis-3-hexenyl salicylate. Results of patch testing and provocative use testing confirmed that she was allergic to octisalate. Provocative use testing indicated that she was also allergic to cis-3-hexenyl salicylate.
Product Description
Supelco
2-Ethylhexyl salicylate, analytical standard
Sigma-Aldrich
2-Ethylhexyl salicylate, 99%
Sigma-Aldrich
2-Ethylhexyl salicylate, ≥99%
Supelco
Octisalate, Pharmaceutical Secondary Standard; Certified Reference Material