Merck
  • Sulfotransferase activities towards xenobiotics and estradiol in two marine fish species (Mullus barbatus and Lepidorhombus boscii): characterization and inhibition by endocrine disrupters.
Sulfotransferase activities towards xenobiotics and estradiol in two marine fish species (Mullus barbatus and Lepidorhombus boscii): characterization and inhibition by endocrine disrupters.
Aquatic toxicology (Amsterdam, Netherlands) (2006-06-30)
Rebeca Martin-Skilton, Michael W H Coughtrie, Cinta Porte
ABSTRACT
We have characterized hepatic phenol sulfotransferase (SULT) activities in two benthic fish species, Mullus barbatus and Lepidorhombus boscii, using p-nitrophenol, dopamine, 17beta-estradiol, 4-nonylphenol, and 1-naphthol as substrates. High affinity sulfation of 17beta-estradiol was observed in both species (Km=28-75 nM), suggesting the presence of a specific estrogen sulfotransferase that catalyzes the formation of estradiol-3 sulfate. Among the tested compounds, 1-naphthol was the most effective substrate for sulfation, with Vmax/Km ratios several hundred-fold higher than the other substrates examined. Both species sulfated the tested compounds, but only M. barbatus was able to sulfate dopamine. We also tested the inhibitory effects of common marine pollutants with estrogenic (4-nonylphenol) and androgenic (tributyltin, triphenyltin) properties on p-nitrophenol and 17beta-estradiol SULT activities. 4-Nonylphenol and triphenyltin inhibited sulfation of both substrates at micromolar concentrations in both species. However, tributyltin was only effective against SULTs from L. boscii, again at micromolar concentrations. The data indicate that M. barbatus and L. boscii are able to sulfate a range of xenobiotics and endogenous compounds, and inhibition of these activities by environmental pollutants may contribute to the known toxic effects of these compounds.
Sigma-Aldrich
β-Estradiol 3-sulfate sodium salt, ≥93%