Merck
  • [Inhibitory effects of phenolic ecotoxicants on photobacteria at various pH values].
[Inhibitory effects of phenolic ecotoxicants on photobacteria at various pH values].
Prikladnaia biokhimiia i mikrobiologiia (2005-12-20)
V V Kuts, Iu M Il'ina, A D Ismailov, A I Netrusov
ABSTRACT
Kinetic characteristics of light emission by intact cells of the photobacteria Photobacterium phosphoreum and Vibrio harveyi at pH 5.5, 7.0, and 8.0 were studied as well as specific features of inhibitory effects of 2,4-di- and 2,4,5-triphenoxyacetic acids (2,4-D and 2,4,5-T), pentachlorophenol (PCP), and 2,6-dimethylphenol (2,6-DMP) at the same pH values. Nonstationarity of emission kinetics was observed at all the pH values studied. Exponential luminescence decay in a 60-sec range was observed at pH 5.5; a 5-min luminescence activation, at pH 7.0 and 8.0. The cell respiratory activity drops by over one order of magnitude at pH 5.5 compared with the activities at pH 7.0 and 8.0. The inhibitory effects of 2,4-D, 2,4,5-T, and PCP differ by one-two orders of magnitude depending on pH. The maximal cell sensitivity to these compounds appears at pH 5.5; the minimal, at pH 8.0. The effect of 2,6-DMP is independent of pH. As is demonstrated, it is hydrophobicity of the molecule and pK values of the toxicants that determine the inhibitory effect. Characteristic of the substrate-starved photobacterial cells are higher sensitivity to chlorophenolic compounds compared with the cells provided with high energy supply at all the pH values.
Sigma-Aldrich
2,6-Dimethylphenol, ≥99.5%
Pricing and availability is not currently available.
Supelco
2,6-Dimethylphenol, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
2,6-Dimethylphenol, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
2,6-Xylenol, ≥99%, FG
Pricing and availability is not currently available.