Merck
  • Structural analysis by reductive cleavage with LiAlH4 of an allyl ether choline-phospholipid, archaetidylcholine, from the hyperthermophilic methanoarchaeon Methanopyrus kandleri.
Structural analysis by reductive cleavage with LiAlH4 of an allyl ether choline-phospholipid, archaetidylcholine, from the hyperthermophilic methanoarchaeon Methanopyrus kandleri.
Archaea (Vancouver, B.C.) (2005-04-05)
Masateru Nishihara, Hiroyuki Morii, Koji Matsuno, Mami Ohga, Karl O Stetter, Yosuke Koga
ABSTRACT
A choline-containing phospholipid (PL-4) in Methanopyrus kandleri cells was identified as archaetidylcholine, which has been described by Sprott et al. (1997). The PL-4 consisted of a variety of molecular species differing in hydrocarbon composition. Most of the PL-4 was acid-labile because of its allyl ether bond. The identity of PL-4 was confirmed by thin-layer chromatography followed by positive staining with Dragendorff reagent and fast-atom bombardment-mass spectrometry. A new method of LiAlH4 hydrogenolysis was developed to cleave allyl ether bonds and recover the corresponding hydrocarbons. We confirmed the validity of the LiAlH4 method in a study of the model compound synthetic unsaturated archaetidic acid (2,3-di-O-geranylgeranyl-sn-glycerol-1-phosphate). Saturated ether bonds were not cleaved by the LiAlH4 method. The hydrocarbons formed following LiAlH4 hydrogenolysis of PL-4 were identified by gas-liquid chromatography and mass spectrometry. Four kinds of hydrocarbons with one to four double bonds were detected: 47% of the hydrocarbons had four double bonds; 11% had three double bonds; 14% had two double bonds; 7% had one double bond; and 6% were saturated species. The molecular species composition of PL-4 was also estimated based on acid lability: 77% of the molecular species had two acid-labile hydrocarbons; 11% had one acid-labile and one acid-stable hydrocarbon; and 11% had two acid-stable hydrocarbons. To our knowledge, this is the first report of a specific chemical degradation method for the structural analysis of allyl ether phospholipid in archaea.
Sigma-Aldrich
Lithium aluminum hydride, hydrogen-storage grade
Sigma-Aldrich
Lithium aluminum hydride solution, 2.0 M in THF
Sigma-Aldrich
Lithium aluminum hydride solution, 2.3 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Lithium aluminum hydride, ≥97.0% (gas-volumetric), tablet (5 g each)
Sigma-Aldrich
Lithium aluminum hydride, ≥97.0% (gas-volumetric)
Sigma-Aldrich
Lithium aluminum hydride solution, 0.5 M in 2-methoxyethyl ether
Sigma-Aldrich
Lithium aluminum hydride solution, 1.0 M in THF
Sigma-Aldrich
Lithium aluminum hydride, pellets, reagent grade, 95%
Sigma-Aldrich
Lithium aluminum hydride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Lithium aluminum hydride, powder, reagent grade, 95%