Merck
Hexazonium pararosaniline as a fixative for animal tissues.
Biotechnic & histochemistry : official publication of the Biological Stain Commission (2005-03-15)
J A Kiernan
ABSTRACT
Hexazonium pararosaniline is a valuable reagent that has been used in enzyme activity histochemistry for 50 years. It is an aqueous solution containing the tris-diazonium ion derived from pararosaniline, an aminotriarylmethane dye, and it contains an excess of nitrous acid that was not consumed in the diazotization reaction. Other investigators have found that immersion for 2 min in an acidic (pH 3.5) 0.0015 M hexazonium pararosaniline solution can protect cryostat sections of unfixed animal tissues from the deleterious effects of aqueous reagents such as buffered solutions used in immunohistochemistry, while preserving specific affinities for antibodies. In the present investigation hexazonium pararosaniline protected lymphoid tissue and striated muscle against the damaging effects of water or saline. The same protection was conferred on unfixed sections treated with dilute nitrous or hydrochloric acid in concentrations similar to those in hexazonium pararosaniline solutions. Model tissues (solutions, gels or films containing gelatin and/or bovine albumin) responded predictably to well known cross-linking (formaldehyde) or coagulant (mercuric chloride) fixatives. Hexazonium pararosaniline solutions prevented the dissolution of protein gels in water only after 9 or more days of contact, during which time considerable swelling occurred. It is concluded that there is no evidence for a "fixative" action of hexazonium pararosaniline. The protective effect on frozen sections of unfixed tissue is attributable probably to the low pH of the solution.
Sigma-Aldrich
Pararosaniline acetate, Dye content 90 %
Sigma-Aldrich
Basic Fuchsin, certified by the Biological Stain Commission, Dye content ≥88 %
Sigma-Aldrich
Basic Fuchsin, Dye content >85 %
Sigma-Aldrich
Pararosaniline hydrochloride, >85.0% (HPLC)