Merck
  • Expression and localization of caveolins during postnatal development in rat heart: implication of thyroid hormone.
Expression and localization of caveolins during postnatal development in rat heart: implication of thyroid hormone.
Journal of applied physiology (Bethesda, Md. : 1985) (2005-02-19)
Philippe Ratajczak, Patricia Oliviéro, Françoise Marotte, Frantisek Kolar, Bohuslav Ostadal, Jane-Lise Samuel
ABSTRACT
Caveolins modulate signaling pathways involved in cardiac development. Caveolin-1 exists in two isoforms: the beta-isoform derivates from an alternative translational start site that creates a protein truncated by 31 amino acids, mainly expressed in endothelial cells, whereas caveolin-3 is present in muscle cells. Our aim was to define caveolin distribution and expression during cardiac postnatal development using immunofluorescence and Western blotting. Caveolin-3 sarcolemmal labeling appeared as dotted lines from days 1 to 5 and as continuous lines after 14 days of age. Caveolin-3 expression, low at birth, increased (4-fold) to reach a maximum (P < 0.05) by day 5 and then decreased to stabilize in adults. Total caveolin-1 and its alpha-isoform were codistributed at birth in endothelial and smooth muscle cells; afterward, only the caveolin-1alpha labeling became limited to endothelium. Quantitative analysis indicated a similar temporal pattern of both total caveolin-1 and caveolin-1alpha expression, suggesting that caveolin-1alpha and -1beta are coregulated; the caveolin-1alpha levels increased fourfold by day 5 to reach a maximum by day 14 (P < 0.05). Tyrosine-14-caveolin-1 phosphorylation, low at birth, increased suddenly around day 14 (8-fold vs. day 1) and returning afterward to basal level. Because the T3/T4 level is maximal by day 14, caveolin-1 expression/phosphorylation profiles were analyzed in hypothyroid heart. The levels of caveolin-1alpha and consequently tyrosine-14-caveolin-1 phosphorylation, but not that of caveolin-3, decreased (50%) in hypothyroid 14-day-old rats. Our data demonstrate that, during postnatal cardiac growth, 1) caveolins are distinctly regulated, and 2) thyroid hormones are involved in caveolin-1alpha expression.
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Supelco
6-Methyl-2-thiouracil, VETRANAL®, analytical standard
Sigma-Aldrich
6-Methyl-2-thiouracil, purum, ≥98.0% S basis (elemental analysis)