Merck
  • Tetramethyl thiourea/Co2(CO)8-catalyzed Pauson-Khand reaction under balloon pressure of CO.
Tetramethyl thiourea/Co2(CO)8-catalyzed Pauson-Khand reaction under balloon pressure of CO.
Organic letters (2005-02-12)
Yefeng Tang, Lujiang Deng, Yandong Zhang, Guangbin Dong, Jiahua Chen, Zhen Yang
ABSTRACT
A Pauson-Khand type of conversion of enynes to bicyclic cyclopentenones employing the commercially available Co2(CO)8 and tetramethylthiourea (TMTU) as catalysts is described. This method allows a variety of enynes with diverse functional groups to be cyclized into cyclopentenones of interest. [reaction: see text]
Sigma-Aldrich
Tetramethylthiourea, 98%