Merck
  • Esterase catalysis of substrate vapour: enzyme activity occurs at very low hydration.
Esterase catalysis of substrate vapour: enzyme activity occurs at very low hydration.
Biochimica et biophysica acta (2004-09-29)
Penelope A Lind, Roy M Daniel, Colin Monk, Rachel V Dunn
ABSTRACT
It has been generally accepted that enzyme activity requires a minimal hydration of about 0.2 g H2O g(-1) protein. This fits well with evidence that hydration above this level is associated with the onset of intramolecular motions. The influence of enzyme hydration on the hydrolysis of substrate by Candida rugosa Lipase B and pig liver esterase was investigated. Each enzyme was studied as a powder at various hydration levels, using vapour phase ethyl butyrate as substrate. This procedure allows the separation of those effects that are due to hydration from those arising from diffusional constraints. We found hydrolytic activity in both enzymes at all hydration levels above zero (between 0.054-0.47 and 0.029-0.60 g H2O g(-1) protein, respectively) that were investigated. The lowest hydration level investigated, <0.03 g H2O g(-1) enzyme, corresponded to a water/enzyme mole ratio of 100 and a coverage of about 10% of the enzyme surface by water molecules. The hydrolytic activity of both enzymes was dependent on protein hydration. However, since the hydrolysis of ethyl butyrate requires water as a second substrate, the absence of activity at zero hydration does not rule out the possibility of enzyme activity in the absence of water. These results suggest that the properties conferred on proteins by water, at least above 10% surface coverage (in this case corresponding to a hydration level of 0.03 g H2O g(-1) protein), are not a requirement for enzyme catalysis.
Product Description
Supelco
Ethyl butyrate, analytical standard
Sigma-Aldrich
Ethyl butyrate, natural, ≥98%, FCC, FG
Sigma-Aldrich
Ethyl butyrate, 99%
Sigma-Aldrich
Ethyl butyrate, ≥98%, FCC, FG