Merck
  • Simulation of organic monolayers as templates for the nucleation of calcite crystals.
Simulation of organic monolayers as templates for the nucleation of calcite crystals.
Langmuir : the ACS journal of surfaces and colloids (2004-08-25)
Dorothy M Duffy, John H Harding
ABSTRACT
Living organisms can control the size, shape, and structure of minerals. Attempts to reproduce this biological control in the laboratory often use Langmuir monolayers of long-chain carboxylic acids. We use large-scale molecular dynamics simulations to calculate the interfacial energies of calcite crystals grown on stearic (octadecanoic) acid monolayers. In light of these simulations we discuss the argument that the orientation of the growing mineral is controlled by the organic substrate acting as a template which the mineral must fit in order to grow.
Product Description
Supelco
Stearyl stearate, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Stearyl stearate, ≥98.0% (GC)
Pricing and availability is not currently available.