Merck
  • Studies on tellurium-containing heterocycles. Part 20. Reactions of 2-benzoselenopyrylium salts and 2-benzotelluropyrylium salts with nucleophiles: formation of 1-functionalized 1H-isoselenochromenes and 1H-isotellurochromenes.
Studies on tellurium-containing heterocycles. Part 20. Reactions of 2-benzoselenopyrylium salts and 2-benzotelluropyrylium salts with nucleophiles: formation of 1-functionalized 1H-isoselenochromenes and 1H-isotellurochromenes.
Chemical & pharmaceutical bulletin (2004-01-08)
Haruki Sashida, Kazuo Ohyanagi
ABSTRACT
The reactions of the 2-benzoselenopyrylium (1A) and 2-benzotelluropyrylium cations (1B) with a variety of nucleophiles have been investigated. LiAlH(4), sodium alkoxide (NaOMe, NaOi-Pr and NaOt-Bu), diethylamine, n-butylamine and acetone reacted with 1 to give the 1H-isochromenes (2) and the corresponding 1-substituted products (4-9) under mild conditions in almost good to high yields. The 1-alkyl(phenyl)isoselenochromenes (10-13) and 1-benzylisochromenes (18A, 18B), which were produced by the reaction of the salts 1 with Grignard reagents, were converted to the corresponding 1,3-disubstituted 2-benzopyrylium salts (14-17, 19) by treatment with triphenylcarbenium tetrafluoroborate (Ph(3)C(+) BF(4)(-)), respectively. The 1-benzylselenopyrylium salts (19A) and 1-benzyltelluropyrylium salts (19B) exist in the solvent as an equilibrium mixture of the salts (19) and the corresponding (Z)-benzylidene compounds (20).
Sigma-Aldrich
Lithium aluminum hydride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Lithium aluminum hydride, ≥97.0% (gas-volumetric)
Sigma-Aldrich
Lithium aluminum hydride solution, 0.5 M in 2-methoxyethyl ether
Sigma-Aldrich
Lithium aluminum hydride, pellets, reagent grade, 95%
Sigma-Aldrich
Lithium aluminum hydride, ≥97.0% (gas-volumetric), tablet (5 g each)
Sigma-Aldrich
Lithium aluminum hydride solution, 1.0 M in THF
Sigma-Aldrich
Lithium aluminum hydride, powder, reagent grade, 95%
Sigma-Aldrich
Lithium aluminum hydride solution, 2.3 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Lithium aluminum hydride solution, 2.0 M in THF
Sigma-Aldrich
Lithium aluminum hydride, hydrogen-storage grade