Merck
  • Skeletal abnormalities in Pth-null mice are influenced by dietary calcium.
Skeletal abnormalities in Pth-null mice are influenced by dietary calcium.
Endocrinology (2004-01-01)
Dengshun Miao, Bin He, Beate Lanske, Xiu-Ying Bai, Xin-Kang Tong, Geoffrey N Hendy, David Goltzman, Andrew C Karaplis
ABSTRACT
We have examined the role of PTH in the postnatal state in a mouse model of PTH deficiency generated by targeting the Pth gene in embryonic stem cells. Mice homozygous for the ablated allele, when maintained on a normal calcium intake, developed hypocalcemia, hyperphosphatemia, and low circulating 1,25-dihydroxyvitamin D(3) [1,25(OH)(2)D(3)] levels consistent with primary hypoparathyroidism. Bone turnover was reduced, leading to increased trabecular and cortical bone volume in PTH-deficient mice. When mutant mice were placed on a low-calcium diet, renal 25-hydroxyvitamin D 1 alpha-hydroxylase expression increased despite the absence of PTH, leading to a rise in circulating 1,25(OH)(2)D(3) levels, marked osteoclastogenesis, and profound bone resorption. These studies demonstrate the dependence of the skeletal phenotype in animals with genetically depleted PTH on the external environment as well as on internal hormonal and ionic circulatory factors. They also show that, although PTH action is the first defense against hypocalcemia, 1,25(OH)(2)D(3) can be mobilized, even in the absence of PTH, to guard against extreme calcium deficiency.
Product Description
Supelco
1α,25-Dihydroxyvitamin D2 solution, 5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
1α,25-Dihydroxyvitamin D2, ≥97.0% (sum of vitamin and previtamin, HPLC)