Merck
  • Determination of 1,3-dichloropropanol in soy and related sauces by using gas chromatography/mass spectrometry.
Determination of 1,3-dichloropropanol in soy and related sauces by using gas chromatography/mass spectrometry.
Food additives and contaminants (2003-11-05)
P J Nyman, G W Diachenko, G A Perfetti
ABSTRACT
A gas chromatography/mass spectrometry method for 3-MCPD in foods and food ingredients was modified for the determination of 1,3-DCP in soy and related sauces. The method was validated by using a blank soy sauce. The detection limit, quantitation limit and recoveries were determined, and identities were confirmed by mass spectrometry on the basis of analyses of test portions spiked with 1,3-DCP at 10, 25, 50 and 100 ng g(-1). The spiked test portions were quantitated by using an internal standard calibration curve. For the spiked test portions, the mean internal standard-corrected recovery for 1,3-DCP was 100% with a relative standard deviation of 1.32%. The limits of detection and quantitation were determined as 0.055 and 0.185 ng g(-1), respectively. The method also was compared with a headspace GC/MS method recently developed by the UK's Central Science Laboratory. Results from the method comparison showed that the recoveries for the spiked test portions, as well as the amounts of 1,3-DCP found in the retail products, were comparable.
Sigma-Aldrich
1,3-Dichloro-2-propanol, 97%
Pricing and availability is not currently available.