Merck
  • Gene cloning and overexpression of two conjugated polyketone reductases, novel aldo-keto reductase family enzymes, of Candida parapsilosis.
Gene cloning and overexpression of two conjugated polyketone reductases, novel aldo-keto reductase family enzymes, of Candida parapsilosis.
Applied microbiology and biotechnology (2003-11-01)
M Kataoka, A-R G Delacruz-Hidalgo, M A Akond, E Sakuradani, K Kita, S Shimizu
ABSTRACT
The genes encoding two conjugated polyketone reductases (CPR-C1, CPR-C2) of Candida parapsilosis IFO 0708 were cloned and sequenced. The genes encoded a total of 304 and 307 amino acid residues for CPR-C1 and CPR-C2, respectively. The deduced amino acid sequences of the two enzymes showed high similarity to each other and to several proteins of the aldo-keto reductase (AKR) superfamily. However, several amino acid residues in putative active sites of AKRs were not conserved in CPR-C1 and CPR-C2. The two CPR genes were overexpressed in Escherichia coli. The E. coli transformant bearing the CPR-C2 gene almost stoichiometrically reduced 30 mg ketopantoyl lactone/ml to D-pantoyl lactone.
Sigma-Aldrich
Dihydro-4,4-dimethyl-2,3-furandione, 97%
Sigma-Aldrich
DL-α-Hydroxy-β,β-dimethyl-γ-butyrolactone, purum, ≥97.0% (T)
Sigma-Aldrich
D-(−)-Pantolactone, 99%
Sigma-Aldrich
(S)-(+)-Pantolactone, 97%