Merck
  • Studies aimed at the total synthesis of azadirachtin. A modeled connection of C-8 and C-14 in azadirachtin.
Studies aimed at the total synthesis of azadirachtin. A modeled connection of C-8 and C-14 in azadirachtin.
Organic letters (2002-08-17)
Takehiro Fukuzaki, Satoshi Kobayashi, Takamasa Hibi, Yohei Ikuma, Jun Ishihara, Naoki Kanoh, Akio Murai
ABSTRACT
[reaction: see text] Studies on the connection between the right and left segments of azadirachtin are described. The Ireland-Claisen rearrangement of Li-enolate of the modeled ester with dichlorodimethylsilane in toluene afforded the desired limonoid framework stereoselectively in good yield.
Sigma-Aldrich
Dichlorodimethylsilane, Wacker Chemie AG, ≥99.0% (GC)
Sigma-Aldrich
Dichlorodimethylsilane, ≥98.5% (GC)
Sigma-Aldrich
Dichlorodimethylsilane, ≥99.5%
Supelco
Silanization solution I, ~5% (dimethyldichlorosilane in heptane), Selectophore