Merck
  • Activation of cellulose membranes with 1,1'-carbonyldiimidazole or 1-cyano-4-dimethylaminopyridinium tetrafluoroborate as a basis for the development of immunosensors.
Activation of cellulose membranes with 1,1'-carbonyldiimidazole or 1-cyano-4-dimethylaminopyridinium tetrafluoroborate as a basis for the development of immunosensors.
Analytical biochemistry (2002-05-16)
Daniela Stöllner, Frieder W Scheller, Axel Warsinke
ABSTRACT
Methods for the activation of a cellulose dialysis membrane for immunosensor applications have been developed. For activation two reagents, 1,1'-carbonyldiimidazole (CDI) and 1-cyano-4-dimethylaminopyridinium tetrafluoroborate (CDAP), were compared with respect to the coupling efficiency for glucose oxidase (GOx) and 1,8-diamino-2,6-dioxaoctane. The maximum level of activation was 2.4 micromol cm(-2) for CDI and 0.2 micromol cm(-2) for CDAP activation. We observed 1.5 microg cm(-2) and 0.4 x 10(-4) U cm(-2) GOx with CDI-activated membranes whereas 1.7 microg cm(-2) and 7.2 x 10(-4) U cm(-2) GOx were observed with CDAP-activated membranes. With 1,8-diamino-2,6-dioxaoctane amino group densities of 0.165 and 0.09 micromol cm(-2) were observed via CDI and CDAP activation, respectively. An amino-modified membrane was used for coupling a ligand (pentapeptide) and an immunoenzymometric assay for hemoglobin A1c was carried out.
Sigma-Aldrich
1-Cyano-4-dimethylaminopyridinium tetrafluoroborate, organic cyanylating reagent