Merck
  • Production, characterization, and cross-reactivity studies of monoclonal antibodies against the coccidiostat nicarbazin.
Production, characterization, and cross-reactivity studies of monoclonal antibodies against the coccidiostat nicarbazin.
Journal of agricultural and food chemistry (2001-10-16)
R C Beier, L H Ripley, C R Young, C M Kaiser
ABSTRACT
A cELISA was developed for the coccidiostat nicarbazin. On the basis of previous computer-assisted molecular modeling studies, p-nitrosuccinanilic acid (PNA-S) was selected as a hapten to produce antibodies to 4,4'-dinitrocarbanilide (DNC), the active component of the coccidiostat nicarbazin. Synthesis is described for the hapten [p-nitro-cis-1,2-cyclohexanedicarboxanilic acid (PNA-C)] used in a BSA conjugate as a plate coating antigen. Monoclonal antibodies (Mabs) were isolated that compete with nicarbazin, having IgM(kappa) isotype. Because of the lack of water solubility of nicarbazin, N,N-dimethylformamide (DMF) (3%, v/v) and acetonitrile (ACN) (10%, v/v) were added to the assay buffer to achieve solubility of nicarbazin and related compounds. The Nic 6 Mabs had an IC(35) value for nicarbazin of 0.92 nmol/mL, with a limit of detection of 0.33 nmol/mL. Nic 6 exhibited high cross-reactivity for PNA-S and PNA-C, and 3-nitrophenol, 4-nitrophenol, and 1-(4-chlorophenyl)-3-(4-nitrophenyl) urea. However, Nic 6 had little or no cross-reactivity with 15 other related compounds.
Sigma-Aldrich
1,3-Bis(4-nitrophenyl)urea, 97%