Merck
  • Juvenile hormone binding protein and transferrin from Galleria mellonella share a similar structural motif.
Juvenile hormone binding protein and transferrin from Galleria mellonella share a similar structural motif.
Biological chemistry (2001-09-04)
D Krzyzanowska, A Ozyhar, A Lalik, J M Parkitna, J Szkudlarek, K Waśniowska, E Lisowska, M Kochman
ABSTRACT
It has been previously suggested that juvenile hormone binding protein(s) (JHBP) belongs to a new class of proteins. In the search for other protein(s) that may contain structural motifs similar to those found in JHBP, hemolymph from Galleria mellonella (Lepidoptera) was chromatographed over a Sephadex G-200 column and resulting fractions were subjected to SDS-PAGE, transferred onto nitrocellulose membrane and scanned with a monoclonal antibody, mAb 104, against hemolymph JHBP. Two proteins yielded a positive reaction with mAb 104, one corresponding to JHBP and the second corresponding to a transferrin, as judged from N-terminal amino acid sequencing staining. Transferrin was purified to about 80% homogeneity using a two-step procedure including Sephadex G-200 gel filtration and HPLC MonoQ column chromatography. Panning of a random peptide display library and analysis with immobilized synthetic peptides were applied for finding a common epitope present in JHBP and the transferrin molecule. The postulated epitope motif recognized by mAb 104 in the JHBP sequence is RDTKAVN, and is localized at position 82-88.
Product Description
Supelco
3-(2-Pyridyl)-5,6-di(2-furyl)-1,2,4-triazine-5′,5′′-disulfonic acid disodium salt, for spectrophotometric det. of Fe, ≥99.0%
Sigma-Aldrich
3-(2-Pyridyl)-5,6-di(2-furyl)-1,2,4-triazine-5′,5′′-disulfonic acid disodium salt, Suitable for determination of Fe