Merck
Synthesis and biological evaluation of destruxin A and related analogs.
The journal of peptide research : official journal of the American Peptide Society (2001-07-17)
T Ast, E Barron, L Kinne, M Schmidt, L Germeroth, K Simmons, H Wenschuh
ABSTRACT
This report describes the development of an efficient solid-phase synthesis protocol and adaptation of reported solution phase procedures for the synthesis of the cyclic depsihexapeptide destruxin A and related analogs. The solid-phase method described is based on standard Fmoc peptide chemistry, including a new synthetic method for the assembly of the depsi bond-containing unit. In order to select analogs of destruxin A for synthesis and evaluation of insecticidal activity, the work of Hellberg et al., describing a set of Z-descriptors for amino acid side-chains comparing their physicochemical properties, was utilized. Destruxin A and 27 different analogs with structural variations in four residues were synthesized and insecticidal activity was evaluated via injections into tobacco budworm (Heliothis virescens) larvae. Several destruxin A analogs were found to be at least as potent as the native compound.
Sigma-Aldrich
Destruxin A from Metarrhizium anisopliae, ≥98% (HPLC)