Merck
  • The current produced by the E779A mutant rat Na(+)/K(+) pump alpha1-subunit expressed in HEK 293 cells.
The current produced by the E779A mutant rat Na(+)/K(+) pump alpha1-subunit expressed in HEK 293 cells.
Biochimica et biophysica acta (2000-12-19)
S Zillikens, G Gisselmann, H G Glitsch
ABSTRACT
The current (I(p)) generated by the wild-type or the glutamate (E) 779 alanine (A) mutant of the rat Na(+)/K(+) pump alpha1-subunit expressed in HEK 293 cells was studied at 35 degrees C by means of whole-cell recording in Na(+)-free and Na(+)-containing solution. Glutamate 779 is located in the fifth transmembrane domain of the alpha-subunit of the Na(+)/K(+)-ATPase. Compared with the wild-type, the E779A mutant exhibited an apparent K(+)(o)-affinity decreased by a factor of 3-4 both in Na(+)-free and in Na(+)-containing media. The competition of Na(+)(o) and K(+)(o) for cation binding sites of the pump remained unchanged. Similarly, in Na(+)-free solution the shape of the I(p)-V curves for various external K(+)-concentrations ([K(+)](o)) was essentially the same. However, in Na(+)-containing solutions the shape of I(p)-V curves from cells expressing the mutant of the rat alpha1-subunit clearly differed from the shape observed in cells expressing the wild-type, but voltage dependence of the pump current persisted. A prominent Na(+)(o)-activated, electrogenic Na(+)-transport mediated by the pump, displaying little voltage dependence in the potential range tested (-80 to +60 mV), was present in the cells expressing the E779A mutant pump. The data suggest that exchanging E779 for A in the rat Na(+)/K(+) pump alpha1-subunit causes a modest decrease in the apparent K(+)(o) affinity and a profound, Na(+)(o)-dependent alteration in the electrogenicity of the mutant pump expressed in HEK 293 cells.
Sigma-Aldrich
Digitoxigenin