MilliporeSigma
  • [Determination of eugenol, methyleugenol and methylisoeugenol in volatile oil of differently processed nutmeg].
[Determination of eugenol, methyleugenol and methylisoeugenol in volatile oil of differently processed nutmeg].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (1997-08-01)
T Jia, M Sha, A Cao, Z Wamg, F Xia
ABSTRACT
A HPLC method for the determination of eugenol, methyleugenol and methylisoeugenol in the volatile oil of differently processed natmeg has been used. The result has shown that the content of eugenol is only sliahtly changed before and after processing, but that of methyleugenol and methylisoeugenol has obviously increased.
Sigma-Aldrich
Methyl isoeugenol, ≥98%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
1,2-Dimethoxy-4-propenylbenzene, 99%
Pricing and availability is not currently available.