Merck
Substance P.
European journal of pain (London, England) (2000-08-25)
D G Snijdelaar, R Dirksen, R Slappendel, B J Crul
ABSTRACT
Substance P is considered to be an important neuropeptide in nociceptive processes. Although substance P was described more than 60 years ago, there is still controversy about its exact role in nociception. This article reviews the current knowledge about the function of substance P in pain. Special emphasis is put on how to use this knowledge in the development of new ways to treat pain.
Sigma-Aldrich
Substance P acetate salt hydrate, ≥95% (HPLC), powder