Merck
  • Isolation of a new gene GS2 (DXS1283E) from a CpG island between STS and KAL1 on Xp22.3.
Isolation of a new gene GS2 (DXS1283E) from a CpG island between STS and KAL1 on Xp22.3.
Genomics (1994-07-15)
W C Lee, E Salido, P H Yen
ABSTRACT
A gene, GS2 (DXS1283E), was isolated from a CpG island located approximately midway between the steroid sulfatase (STS) and the Kallmann syndrome (KAL1) loci on the distal short arm of the human X chromosome. DNA sequencing of a GS2 cDNA clone revealed an open reading frame for a basic protein of 253 amino acid residues. A polymorphic CT dinucleotide repeat was found in the 3' untranslated region. The GS2 gene is expressed in all human tissues examined, including heart, brain, placenta, lung, liver, muscle, kidney, pancreas, and spleen. Several GS2 transcripts, ranging in size from 1.1 to 5.8 kb, were found among different tissues, suggesting tissue-specific processing of the GS2 transcript. Characterization of GS2 genomic clones revealed that the gene consists of 7 exons spanning over 26 kb, with a CpG island located in the first intron. The GS2 gene is transcribed toward Xpter, in the same direction as KAL1 but opposite to that of STS.
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg