Merck
  • Poly(hydroxyalkanoates)-based polymeric nanoparticles for drug delivery.
Poly(hydroxyalkanoates)-based polymeric nanoparticles for drug delivery.
Journal of biomedicine & biotechnology (2009-10-01)
Cesare Errico, Cristina Bartoli, Federica Chiellini, Emo Chiellini
ABSTRACT
Poly (hydroxyalkanoates) (PHAs) have recently attracted a great deal of academic and industrial interest for their biodegradability and biocompatibility making them suitable for environmental and biomedical applications. Poly(3-hydroxybutyrate-) (PHB-) and Poly(DL-lactide-co-glycolide) (PLGA-) based nanoparticles were prepared using the dialysis method as yet unreported for the preparation of nanoparticles based on PHB. Processing conditions were varied in order to evaluate their influence on morphology, drug encapsulation, and size of nanoparticles. The relevant results obtained give a theoretical understanding of the phenomenon occurring during colloidal formation. The adopted procedure allows for a relatively small diameter and homogeneity in size distribution of the PHB nanoparticles to be obtained compared to other methods like the one based on solvent evaporation which leads to particles on microscale. The biocompatibility of PHB and relative nanoparticles was investigated and both exhibited very good cytocompatibility.
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications