- Isoguvacine, isonipecotic acid, muscimol and N-methyl isoguvacine on the GABA receptor in rat sympathetic ganglia.
Isoguvacine, isonipecotic acid, muscimol and N-methyl isoguvacine on the GABA receptor in rat sympathetic ganglia.
The GABA-mimetic activities of 4 analogues muscimol, isonipecotic acid, isoguvacine and N-methyl isoguvacine have been examined at the GABA receptor in the rat isolated superior cervical ganglion. The depolarizing action of all 4 analogues could be selectively antagonized by bicuculline methochloride and isopropyl bicyclophosphate. Muscimol was the only analogue more potent than GABA (molar potency ratio = 5.08 +/- 0.707). The potency of isoguvacine was 0.23 +/- 0.026 and isonipecotic acid 0.011 +/- 0.0028. N-methyl isoguvacine was less than 0.001 GABA.