Merck
  • Identification of the active form of endothelial lipase, a homodimer in a head-to-tail conformation.
Identification of the active form of endothelial lipase, a homodimer in a head-to-tail conformation.
The Journal of biological chemistry (2009-07-02)
Nathalie Griffon, Weijin Jin, Thomas J Petty, John Millar, Karen O Badellino, Jeffery G Saven, Dawn H Marchadier, Ellis S Kempner, Jeffrey Billheimer, Jane M Glick, Daniel J Rader
ABSTRACT
Endothelial lipase (EL) is a member of a subfamily of lipases that act on triglycerides and phospholipids in plasma lipoproteins, which also includes lipoprotein lipase and hepatic lipase. EL has a tropism for high density lipoprotein, and its level of phospholipase activity is similar to its level of triglyceride lipase activity. Inhibition or loss-of-function of EL in mice results in an increase in high density lipoprotein cholesterol, making it a potential therapeutic target. Although hepatic lipase and lipoprotein lipase have been shown to function as homodimers, the active form of EL is not known. In these studies, the size and conformation of the active form of EL were determined. Immunoprecipitation experiments suggested oligomerization. Ultracentrifugation experiments showed that the active form of EL had a molecular weight higher than the molecular weight of a simple monomer but less than a dimer. A construct encoding a covalent head-to-tail homodimer of EL (EL-EL) was expressed and had similar lipolytic activity to EL. The functional molecular weights determined by radiation inactivation were similar for EL and the covalent homodimer EL-EL. We previously showed that EL could be cleaved by proprotein convertases, such as PC5, resulting in loss of activity. In cells overexpressing PC5, the covalent homodimeric EL-EL appeared to be more stable, with reduced cleavage and conserved lipolytic activity. A comparative model obtained using other lipase structures suggests a structure for the head-to-tail EL homodimer that is consistent with the experimental findings. These data confirm the hypothesis that EL is active as a homodimer in head-to-tail conformation.
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)