Merck
  • Enhanced release of IgE-dependent early phase mediators from nasal polyp tissue.
Enhanced release of IgE-dependent early phase mediators from nasal polyp tissue.
Journal of inflammation (London, England) (2009-04-22)
Joke Patou, Gabriele Holtappels, Karen Affleck, Philippe Gevaert, Claudina Perez-Novo, Paul Van Cauwenberge, Claus Bachert
ABSTRACT
The mast cell is a crucial effector cell in allergic rhinitis and other inflammatory diseases. During the acute allergic reaction preformed mediators such as histamine, but also de novo produced mediators such as leukotrienes (LTC4/D4/E4) and prostaglandins (PGD2) are released. Mast cells represent targets for therapeutic intervention, and thus a human ex-vivo model to stimulate mast cells taken from mucosal sites would be instrumental for drug intervention studies. We have aimed to activate mast cells within ex-vivo human nasal tissue by IgE/anti-IgE specific (epsilon chain specific) stimulations and in this respect to test the usability of nasal polyps versus inferior turbinates Biopsy samples were collected from patients with nasal polyps and inferior turbinates from patients who underwent sinus or septal surgery. Tissue fragments were primed with IgE 1 mug/ml for 60 minutes and then stimulated for 30 minutes with tissue culture medium (negative control), anti-IgE 10 mug/ml, anti-IgE 30 mug/ml and ionomycin 10 muM (positive control). Histamine, leukotrienes and PGD2 were measured in supernatants. To help provide an understanding of the extent of the response, the number of tryptase and FcepsilonRIalpha positive cells was evaluated by means of immunohistochemistry and the FcepsilonRIalpha-chain was measured by means of quantitative PCR in the nasal polyp and inferior turbinate tissues. Finally, the correlation between IgE concentrations in the nasal tissue and the release of mediators was analysed. Stimulations with anti-IgE on IgE-primed nasal tissue fragments lead to a concentration-dependent release of histamine, leukotrienes and PGD2. The release of these early phase mediators was significantly higher in nasal polyps compared to inferior turbinates, although tryptase, FcepsilonRIalpha positive cells and FcepsilonRIalpha-chain transcripts were equally present in both groups. No correlation was found between baseline concentrations of IgE, and the release of histamine, LTC4/LTD4/LTE4 and PGD2 after stimulation. This human nasal challenge model mimics the allergic early phase reaction. The release of histamine, cys-leukotrienes and PGD2 was significantly higher in nasal polyps versus inferior turbinates, however, this observation could not be explained by differences in mast cell or FcepsilonRI+ cell numbers.
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate. Without arginine, leucine, lysine, and phenol red, liquid, sterile-filtered, suitable for cell culture, designed for isotope labeling for cell culture applications
Sigma-Aldrich
StableCell RPMI-1640, With stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, with 2.05 mM L-glutamine, with 25mM HEPES, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, 10 ×, Without L-glutamine, folic acid and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with 20 mM HEPES and L-glutamine, without sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with L-glutamine, without phenol red and sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640 Medium, HEPES Modification, with L-glutamine and 25mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without methionine, cystine and L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Dutch Modification, with sodium bicarbonate and 20mM HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, AutoMod, without L-glutamine and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without glucose and sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640 Medium, Hybri-Max, Modified, with L-glutamine, 4500 mg/L glucose and 15mM HEPES, without sodium bicarbonate, powder, suitable for hybridoma
Sigma-Aldrich
RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture