Merck
  • Regulation of eumelanin/pheomelanin synthesis and visible pigmentation in melanocytes by ligands of the melanocortin 1 receptor.
Regulation of eumelanin/pheomelanin synthesis and visible pigmentation in melanocytes by ligands of the melanocortin 1 receptor.
Pigment cell & melanoma research (2008-07-17)
Elodie Le Pape, Kazumasa Wakamatsu, Shosuke Ito, Rainer Wolber, Vincent J Hearing
ABSTRACT
The production of melanin in the hair and skin is tightly regulated by the melanocortin 1 receptor (MC1R) whose activation is controlled by two secreted ligands, alpha-melanocyte stimulating hormone (alphaMSH) and agouti signal protein (ASP). As melanin is extremely stable, lasting years in biological tissues, the mechanism underlying the relatively rapid decrease in visible pigmentation elicited by ASP is of obvious interest. In this study, the effects of ASP and alphaMSH on the regulation of melanin synthesis and on visible pigmentation were assessed in normal murine melanocytes and were compared with the quick depigmenting effect of the tyrosinase inhibitor, phenylthiourea (PTU). alphaMSH increased pheomelanin levels prior to increasing eumelanin content over 4 days of treatment. Conversely, ASP switched off the pigment synthesis pathway, reducing eu- and pheo-melanin synthesis within 1 day of treatment that was proportional to the decrease in tyrosinase protein level and activity. These results demonstrate that the visible depigmentation of melanocytes induced by ASP does not require the degradation of existing melanin but rather is due to the dilution of existing melanin by melanocyte turnover, which emphasizes the importance of pigment distribution to visible color.
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications