Merck
  • Cloning of a unique lipase from endothelial cells extends the lipase gene family.
Cloning of a unique lipase from endothelial cells extends the lipase gene family.
The Journal of biological chemistry (1999-05-13)
K Hirata, H L Dichek, J A Cioffi, S Y Choi, N J Leeper, L Quintana, G S Kronmal, A D Cooper, T Quertermous
ABSTRACT
A new lipoprotein lipase-like gene has been cloned from endothelial cells through a subtraction methodology aimed at characterizing genes that are expressed with in vitro differentiation of this cell type. The conceptual endothelial cell-derived lipase protein contains 500 amino acids, including an 18-amino acid hydrophobic signal sequence, and is 44% identical to lipoprotein lipase and 41% identical to hepatic lipase. Comparison of primary sequence to that of lipoprotein and hepatic lipase reveals conservation of the serine, aspartic acid, and histidine catalytic residues as well as the 10 cysteine residues involved in disulfide bond formation. Expression was identified in cultured human umbilical vein endothelial cells, human coronary artery endothelial cells, and murine endothelial-like yolk sac cells by Northern blot. In addition, Northern blot and in situ hybridization analysis revealed expression of the endothelial-derived lipase in placenta, liver, lung, ovary, thyroid gland, and testis. A c-Myc-tagged protein secreted from transfected COS7 cells had phospholipase A1 activity but no triglyceride lipase activity. Its tissue-restricted pattern of expression and its ability to be expressed by endothelial cells, suggests that endothelial cell-derived lipase may have unique functions in lipoprotein metabolism and in vascular disease.
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg