Merck
Interaction between FXIII and fibrinogen.
Blood (2013-03-16)
Hans P Kohler
ABSTRACT
In this issue of Blood, Smith and colleagues report on the functional role of the interaction between these 2 proteins by studying the involved binding sites responsible for clot stabilization.(1)
Sigma-Aldrich
Thrombin from bovine plasma, ≥60 NIH units/mg protein (biuret)
Sigma-Aldrich
Thrombin from bovine plasma, lyophilized powder, 600-2,000 NIH units/mg protein (biuret)
Sigma-Aldrich
Thrombin from bovine plasma, lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280 = 19.5)
Sigma-Aldrich
Thrombin from bovine plasma, lyophilized powder, 40-500 NIH units/mg protein (biuret)
Sigma-Aldrich
Thrombin human, BioUltra, recombinant, expressed in HEK 293 cells, aqueous solution, ≥95% (SDS-PAGE)
Sigma-Aldrich
Thrombin from human plasma, 400-1000 NIH units/mg protein
Sigma-Aldrich
Thrombin from human plasma, lyophilized powder, Suitable for routine use in the thrombin time test
Sigma-Aldrich
Thrombin from human plasma, lyophilized powder, 1500-3500 NIH units/mg protein (E1%/280, 18.3), suitable for cell culture
Sigma-Aldrich
Thrombin from human plasma, lyophilized powder, ≥2800 NIH units/mg protein (E1%/280, 18.3)
Sigma-Aldrich
Thrombin from human plasma, lyophilized powder, ≥1,000 NIH units/mg protein (E1%/280, 18.3)
Sigma-Aldrich
Thrombin from human plasma, lyophilized powder, ≥2,000 NIH units/mg protein (E1%/280, 18.3)