Merck
  • High-fat diet-mediated lipotoxicity and insulin resistance is related to impaired lipase expression in mouse skeletal muscle.
High-fat diet-mediated lipotoxicity and insulin resistance is related to impaired lipase expression in mouse skeletal muscle.
Endocrinology (2013-03-09)
Pierre-Marie Badin, Isabelle K Vila, Katie Louche, Aline Mairal, Marie-Adeline Marques, Virginie Bourlier, Geneviève Tavernier, Dominique Langin, Cedric Moro
ABSTRACT
Elevated expression/activity of adipose triglyceride lipase (ATGL) and/or reduced activity of hormone-sensitive lipase (HSL) in skeletal muscle are causally linked to insulin resistance in vitro. We investigated here the effect of high-fat feeding on skeletal muscle lipolytic proteins, lipotoxicity, and insulin signaling in vivo. Five-week-old C3H mice were fed normal chow diet (NCD) or 45% kcal high-fat diet (HFD) for 4 weeks. Wild-type and HSL knockout mice fed NCD were also studied. Whole-body and muscle insulin sensitivity, as well as lipolytic protein expression, lipid levels, and insulin signaling in skeletal muscle, were measured. HFD induced whole-body insulin resistance and glucose intolerance and reduced skeletal muscle glucose uptake compared with NCD. HFD increased skeletal muscle total diacylglycerol (DAG) content, protein kinase Cθ and protein kinase Cε membrane translocation, and impaired insulin signaling as reflected by a robust increase of basal Ser1101 insulin receptor substrate 1 phosphorylation (2.8-fold, P < .05) and a decrease of insulin-stimulated v-Akt murine thymoma viral oncogene homolog Ser473 (-37%, P < .05) and AS160 Thr642 (-47%, P <.01) phosphorylation. We next showed that HFD strongly reduced HSL phosphorylation at Ser660. HFD significantly up-regulated the muscle protein content of the ATGL coactivator comparative gene identification 58 and triacylglycerol hydrolase activity, despite a lower ATGL protein content. We further show a defective skeletal muscle insulin signaling and DAG accumulation in HSL knockout compared with wild-type mice. Together, these data suggest a pathophysiological link between altered skeletal muscle lipase expression and DAG-mediated insulin resistance in mice.
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥10 U/mg
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Esterase from porcine liver, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Esterase from rabbit liver, lyophilized powder, ≥30 units/mg protein
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lipase from Aspergillus oryzae, solution, ≥100,000 U/g, white, beige
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Esterase from porcine liver, lyophilized, powder, slightly beige, ≥50 U/mg
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Esterase Isoenzyme 1 porcine liver, recombinant, recombinant, expressed in E. coli, ≥30.0 U/g
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, ≥0.2 U/mg
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg