Merck
  • Polar residues and their positional context dictate the transmembrane domain interactions of influenza A neuraminidases.
Polar residues and their positional context dictate the transmembrane domain interactions of influenza A neuraminidases.
The Journal of biological chemistry (2013-03-01)
Johan Nordholm, Diogo V da Silva, Justina Damjanovic, Dan Dou, Robert Daniels
ABSTRACT
Interactions that facilitate transmembrane domain (TMD) dimerization have been identified mainly using synthetic TMDs. Here, we investigated how inherent properties within natural TMDs modulate their interaction strength by exploiting the sequence variation in the nine neuraminidase subtypes (N1-N9) and the prior knowledge that a N1 TMD oligomerizes. Initially, consensus TMDs were created from the influenza A virus database, and their interaction strengths were measured in a biological membrane system. The TMD interactions increased with respect to decreasing hydrophobicity across the subtypes (N1-N9) and within the human N1 subtype where the N1 TMDs from the pandemic H1N1 strain of swine origin were found to be significantly less hydrophobic. The hydrophobicity correlation was attributed to the conserved amphipathicity within the TMDs as the interactions were abolished by mutating residues on the polar faces that are unfavorably positioned in the membrane. Similarly, local changes enhanced the interactions only when a larger polar residue existed on the appropriate face in an unfavorable membrane position. Together, the analysis of this unique natural TMD data set demonstrates how polar-mediated TMD interactions from bitopic proteins depend on which polar residues are involved and their positioning with respect to the helix and the membrane bilayer.
Sigma-Aldrich
α(2→3,6) Neuraminidase from Clostridium perfringens (C. welchii), recombinant, expressed in E. coli, buffered aqueous solution, ≥250 units/mg protein
Sigma-Aldrich
Neuraminidase Agarose from Clostridium perfringens (C. welchii), Type VI-A, ammonium sulfate suspension
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VIII, lyophilized powder, 10-20 units/mg protein (using 4MU-NANA), 3.5-8.0 units/mg protein (mucin)
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VI, lyophilized powder, 6-15 units/mg protein (using 4MU-NANA), 2-10 units/mg protein (mucin)
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type X, lyophilized powder, ≥50 units/mg protein (using 4MU-NANA)
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Suitable for manufacturing of diagnostic kits and reagents, Type V, lyophilized powder
Sigma-Aldrich
α(2→3) Neuraminidase from Streptococcus pneumoniae, buffered aqueous solution
Sigma-Aldrich
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, Proteomics Grade, suitable for MALDI-TOF MS
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, ≥1.5 U/mL, specific activity ≥ 1.5U/mg protein
Sigma-Aldrich
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, Type III, buffered aqueous solution, 0.2 μm filtered, 1-5 units/mg protein (Lowry, using NAN-lactose)
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, Type II, buffered aqueous solution, 8-24 units/mg protein (Lowry, using NAN-lactose)