Merck
  • A universal, high recovery assay for protein quantitation through temperature programmed liquid chromatography (TPLC).
A universal, high recovery assay for protein quantitation through temperature programmed liquid chromatography (TPLC).
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2013-02-26)
Dennis J Orton, Alan A Doucette
ABSTRACT
As an alternative to direct UV absorbance measurements, estimation of total protein concentration is typically conducted through colorimetric reagent assays. However, for protein-limited applications, the proportion of the sample sacrificed to the assay becomes increasingly significant. This work demonstrates a method for quantitation of protein samples with high recovery. Temperature programmed liquid chromatography (TPLC) with absorbance detection at 214nm permits accurate estimation of total protein concentration from samples containing as little as 0.75μg. The method incorporates a temperature gradient from 25 to 80°C to facilitate elution of total protein into a single fraction. Analyte recovery, as measured from 1 and 10μg protein extracts of Escherichia coli, is shown to exceed 93%. Extinction coefficients at 214nm were calculated across the human proteome, providing a relative standard deviation of 21% (versus 42% at 280nm), suggesting absorbance values at 214nm provide a more consistent measure of protein concentration. These results translate to a universal protein detection strategy exhibiting a coefficient of variation below 10%. Together with the sensitivity and tolerance to contaminants, TPLC with UV detection is a favorable alternative to colorimetric assay for total protein quantitation, particularly in sample-limited applications.
Sigma-Aldrich
Lysozyme from human neutrophils, ≥95% (SDS-PAGE), lyophilized powder, ≥100,000 units/mg protein (E1%/280)
Sigma-Aldrich
Lysozyme human, Lysobac, recombinant, expressed in rice, ≥100,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lysozyme from chicken egg white, BioUltra, lyophilized powder, ≥98% (SDS-PAGE), ≥40,000 units/mg protein
Supelco
Lysozyme from chicken egg white, VETRANAL®, analytical standard
Sigma-Aldrich
Lysozyme from chicken egg white, dialyzed, lyophilized, powder, ~100000 U/mg
Sigma-Aldrich
Lysozyme from chicken egg white, powder (crystalline), 70000-140000 U/mg
Sigma-Aldrich
Lysozyme from chicken egg white, 10 mg/mL
Sigma-Aldrich
Lysozyme chloride form from chicken egg white, Grade VI, ≥35,000 units/mg protein (E1%/282)
Sigma-Aldrich
Lysozyme from chicken egg white, For use as a marker in SDS-PAGE
Sigma-Aldrich
Lysozyme from chicken egg white, aseptically filled
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥90 %, ≥39,000 units/mg protein