Merck
  • Reciprocal regulation of pH 6 antigen gene loci by PhoP and RovA in Yersinia pestis biovar Microtus.
Reciprocal regulation of pH 6 antigen gene loci by PhoP and RovA in Yersinia pestis biovar Microtus.
Future microbiology (2013-02-05)
Yiquan Zhang, Li Wang, Nan Fang, Shi Qu, Yafang Tan, Zhaobiao Guo, Jingfu Qiu, Dongsheng Zhou, Ruifu Yang
ABSTRACT
To explore the transcriptional regulation of the psaEF and psaABC loci by the RovA and PhoP regulators in Yersinia pestis. Primer extension, LacZ fusion, gel mobility shift and DNase I footprinting assays were conducted in combination for this gene regulation study. It was determined that PhoP and RovA recognized the promoter-proximal regions of psaEF and psaABC in order to repress and stimulate their transcription, respectively. The translation/transcription start sites, Shine-Dalgarno sequences (ribosomal binding site), core promoter -10 and -35 elements, PhoP and RovA sites and PhoP/RovA consensus-like sequences were identified to determine the structural organization of PhoP/RovA-dependent promoters of psaEF and psaABC. RovA stimulated psaEF and psaABC, while PhoP repressed these two operons involving the direct association between RovA/PhoP and target promoter regions. The reciprocal regulation of psa genes by PhoP and RovA could contribute to the tightly controlled expression of the pH 6 antigen during infection.
Sigma-Aldrich
β-Galactosidase from Aspergillus oryzae, ≥8.0 units/mg solid
Sigma-Aldrich
β-Galactosidase from Escherichia coli, suitable for enzyme immunoassay, lyophilized, powder, ~140 U/mg
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VI, lyophilized powder, ≥250 units/mg protein
Sigma-Aldrich
β-Galactosidase from bovine liver, Grade III, lyophilized powder, ≥0.15 units/mg protein
Sigma-Aldrich
β-Galactosidase from Escherichia coli, lyophilized powder, ≥500 units/mg protein
Sigma-Aldrich
β-Galactosidase from bovine testes, ammonium sulfate suspension, 1.0-3.0 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
β-Galactosidase from Escherichia coli, aqueous glycerol suspension, ≥500 units/mg protein (biuret)
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VIII, lyophilized powder, ≥500 units/mg protein