Merck
  • Structure-based design and synthesis of C-1- and C-4-modified analogs of zanamivir as neuraminidase inhibitors.
Structure-based design and synthesis of C-1- and C-4-modified analogs of zanamivir as neuraminidase inhibitors.
Journal of medicinal chemistry (2013-01-22)
Enguang Feng, Woo-Jin Shin, Xuelian Zhu, Jian Li, Deju Ye, Jiang Wang, Mingyue Zheng, Jian-Ping Zuo, Kyoung Tai No, Xian Liu, Weiliang Zhu, Wei Tang, Baik-Lin Seong, Hualiang Jiang, Hong Liu
ABSTRACT
In order to exploit the 430-cavity in the active sites of neuraminidases, 22 zanamivir analogs with C-1 and C-4 modification were synthesized, and their inhibitory activities against both group-1 (H5N1, H1N1) and group-2 neuraminidases (H3N2) were determined. Compound 9f exerts the most potency, with IC(50) value of 0.013, 0.001, and 0.09 μM against H3N2, H5N1, and H1N1, which is similar to that of zanamivir (H3N2 IC(50) = 0.0014 μM, H5N1 IC(50) = 0.012 μM, H1N1 IC(50) = 0.001 μM). Pharmacokinetic studies of compound 9f in rats showed a much longer plasma half-life (t(1/2)) than that of zanamivir following administration (po dose). Molecular modeling provided information about the binding model between the new inhibitors and neuraminidase, with the elongated groups at the C-1-position being projected toward the 430-loop region. This study may represent a novel starting point for the future development of improved antiflu agents.
Sigma-Aldrich
Neuraminidase Agarose from Clostridium perfringens (C. welchii), Type VI-A, ammonium sulfate suspension
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VI, lyophilized powder, 6-15 units/mg protein (using 4MU-NANA), 2-10 units/mg protein (mucin)
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type X, lyophilized powder, ≥50 units/mg protein (using 4MU-NANA)
Sigma-Aldrich
α(2→3) Neuraminidase from Streptococcus pneumoniae, buffered aqueous solution
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Suitable for manufacturing of diagnostic kits and reagents, Type V, lyophilized powder
Sigma-Aldrich
α(2→3,6) Neuraminidase from Clostridium perfringens (C. welchii), recombinant, expressed in E. coli, buffered aqueous solution, ≥250 units/mg protein
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VIII, lyophilized powder, 10-20 units/mg protein (using 4MU-NANA), 3.5-8.0 units/mg protein (mucin)
Sigma-Aldrich
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, Type III, buffered aqueous solution, 0.2 μm filtered, 1-5 units/mg protein (Lowry, using NAN-lactose)
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, Type II, buffered aqueous solution, 8-24 units/mg protein (Lowry, using NAN-lactose)
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, ≥1.5 U/mL, specific activity ≥ 1.5U/mg protein
Sigma-Aldrich
Zanamivir, ≥98% (HPLC)
Sigma-Aldrich
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, Proteomics Grade, suitable for MALDI-TOF MS