Merck
  • Comprehensive profiling of N-linked glycosylation sites in HeLa cells using hydrazide enrichment.
Comprehensive profiling of N-linked glycosylation sites in HeLa cells using hydrazide enrichment.
Journal of proteome research (2012-12-05)
Helle Malerod, Robert L J Graham, Michael J Sweredoski, Sonja Hess
ABSTRACT
The adenocarcinoma cell line HeLa serves as a model system for cancer research in general and cervical cancer in particular. In this study, hydrazide enrichment in combination with state-of-the art nanoLC-MS/MS analysis was used to profile N-linked glycosites in HeLa cells. N-Linked glycoproteins were selectively enriched in HeLa cells by the hydrazide capture method, which isolates all glycoproteins independent of their glycans. Nonglycosylated proteins were removed by extensive washing. N-Linked glycoproteins were identified with the specific NXT/S motif and deamidated asparagine (N). Deglycosylation was carried out in both H(2)(16)O and H(2)(18)O to confirm the deamidation. NanoLC-MS/MS analysis indicated that the method selectively enriched at least 100 fold N-linked glycosites in HeLa cells. When both the membrane and cytosolic fractions were used, a total of 268 unique N-glycosylation sites were identified corresponding to 106 glycoproteins. Bioinformatic analysis revealed that most of the glycoproteins identified are known to have an impact on cancer and have been proposed as biomarkers.
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, recombinant, expressed in E. coli, set of 100 units nanomolar unit
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, BioReagent, ≥95% (SDS-PAGE)
Sigma-Aldrich
Glycopeptidase A from almonds, buffered aqueous glycerol solution, ≥0.05 unit/mL
Sigma-Aldrich
PNGase F from Elizabethkingia miricola, buffered aqueous solution
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, lyophilized powder, recombinant, expressed in E. coli
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, ready-to-use solution, recombinant, expressed in E. coli