Merck
  • Site-selective azide incorporation into endogenous RNase A via a "chemistry" approach.
Site-selective azide incorporation into endogenous RNase A via a "chemistry" approach.
Organic & biomolecular chemistry (2012-11-23)
Xi Chen, Lars Henschke, Qianzhen Wu, Kasturi Muthoosamy, Boris Neumann, Tanja Weil
ABSTRACT
Site-selective labeling of endogenous proteins represents a major challenge in chemical biology, mainly due to the absence of unique reactive groups that can be addressed selectively. Recently, we have shown that surface-exposed lysine residues of two endogenous proteins and a peptide exhibit subtle changes in their individual reactivities. This feature allows the modification of a single residue in a highly site-selective fashion if kinetically controlled labeling conditions are applied. In order to broaden the scope of the "kinetically-controlled protein labeling" (KPL) approach and highlight additional applications, the water-soluble bioorthogonal reagent, biotin-TEO-azido-NHS (11), is developed which enables the site-selective introduction of an azido group onto endogenous proteins/peptides. This bioconjugation reagent features a biotin tag for affinity purification, an azido group for bioorthogonal labeling, a TEO (tetraethylene oxide) linker acting as a spacer and to impart water solubility and an N-hydroxysuccinimidyl (NHS) ester group for reacting with the exposed lysine residue. As a proof of concept, the native protein ribonuclease A (RNase A) bearing ten available lysine residues at the surface is furnished with a single azido group at Lys 1 in a highly site-selective fashion yielding azido-(K1)RNase A. The K1 site-selectivity is demonstrated by the combined application and interpretation of high resolution MALDI-ToF mass spectroscopy, tandem mass spectroscopy and extracted ion chromatography (XIC). Finally, the water soluble azide-reactive phosphine probe, rho-TEO-phosphine (21) (rho: rhodamine), has been designed and applied to attach a chromophore to azido-(K1)RNase A via Staudinger ligation at physiological pH indicating that the introduced azido group is accessible and could be addressed by other established azide-reactive bioorthogonal reaction schemes.
Sigma-Aldrich
Ethylene oxide, ≥99.5%
Supelco
Ethylene oxide solution, certified reference material, 50 mg/mL in methanol
Sigma-Aldrich
Ethylene oxide solution, 2.5-3.3 M in THF
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-AS, 50-100 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type III-A, ≥85% RNase A basis (SDS-PAGE), 85-140 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type II-A, ≥60% (SDS-PAGE), >= 60 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type X-A, ≥90% (SDS-PAGE), ≥70 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, (Solution of 50% glycerol, 10mM Tris-HCL pH 8.0)
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type XII-A, ≥90% (SDS-PAGE), 75-125 Kunitz units/mg protein
Sigma-Aldrich
Ribonuclease A from bovine pancreas, for molecular biology, ≥70 Kunitz units/mg protein, lyophilized
Sigma-Aldrich
Ribonuclease B from bovine pancreas, BioReagent, ≥50 Kunitz units/mg protein, ≥80% (SDS-PAGE)
Sigma-Aldrich
Ribonuclease A-agarose, ammonium sulfate suspension, 400-1,000 units/g agarose (One ml gel will yield 12-30 units)