Merck
  • Invertase SUC2 Is the key hydrolase for inulin degradation in Saccharomyces cerevisiae.
Invertase SUC2 Is the key hydrolase for inulin degradation in Saccharomyces cerevisiae.
Applied and environmental microbiology (2012-10-30)
Shi-An Wang, Fu-Li Li
ABSTRACT
Specific Saccharomyces cerevisiae strains were recently found to be capable of efficiently utilizing inulin, but genetic mechanisms of inulin hydrolysis in yeast remain unknown. Here we report functional characteristics of invertase SUC2 from strain JZ1C and demonstrate that SUC2 is the key enzyme responsible for inulin metabolism in S. cerevisiae.
Sigma-Aldrich
Invertase from baker′s yeast (S. cerevisiae), Grade VII, ≥300 units/mg solid
Supelco
Invertase from baker′s yeast (S. cerevisiae), 200-300 units/mg solid
Sigma-Aldrich
Invertase from Candida utilis, Grade X, ≥300 units/mg solid
Sigma-Aldrich
Invertase Glycoprotein Standard, BioReagent, from Saccharomyces cerevisiae