Merck
  • Structural changes and allergenic properties of β-lactoglobulin upon exposure to high-intensity ultrasound.
Structural changes and allergenic properties of β-lactoglobulin upon exposure to high-intensity ultrasound.
Molecular nutrition & food research (2012-10-16)
Dragana Stanic-Vucinic, Marija Stojadinovic, Marina Atanaskovic-Markovic, Jana Ognjenovic, Hans Grönlund, Marianne van Hage, Raija Lantto, Ana I Sancho, Tanja Cirkovic Velickovic
ABSTRACT
The aim of this work was to investigate the effects of high-intensity ultrasound (sonication), on the structure and allergenicity of the major cow's milk allergen, beta-lactoglobulin (BLG). Structural changes upon sonication of BLG were monitored by circular dichroism spectroscopy, tryptophan emission fluorescence, hydrophobic dye and retinol binding, as well as digestibility and phenol-oxidase cross-linking capacity. Allergenicity was monitored in individual patients' sera, basophil activation test, and skin prick testing in 41 cow's milk allergy patients. Uncontrolled local temperature changes induced modifications in BLG secondary structure accompanied by formation of dimers, trimers, and oligomers of BLG that were more digestible by pepsin and had reduced retinol binding. Controlled temperature conditions induced changes in secondary structure of BLG without causing formation of oligomers, or changing protein's capacity to bind retinol. Both sonicated forms of BLG had more exposed hydrophobic surfaces than native BLG and underwent facilitated cross-linking reaction with phenol-oxidase. Sonication had a minor effect on IgE-binding properties of BLG. Sonication-induced structural changes in major whey allergen were not clinically significant in cow's milk allergy patients. Ultrasound can be a safe procedure for dairy processing as it maintains the nutritional value and does not increase allergenic potential of BLG.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, tested according to Ph. Eur.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, slightly beige, ≥500 U/mg
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, slightly beige, 1200-2400 U/mg
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥400 units/mg protein
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥2,500 units/mg protein (E1%/280)
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Sigma-Aldrich
Pepsin−Agarose from porcine gastric mucosa, lyophilized powder, ≥30 units/mg dry solid