Merck
  • Increased red cell turnover in a line of CD22-deficient mice is caused by Gpi1c: a model for hereditary haemolytic anaemia.
Increased red cell turnover in a line of CD22-deficient mice is caused by Gpi1c: a model for hereditary haemolytic anaemia.
European journal of immunology (2012-08-30)
Jennifer A Walker, Andrew M Hall, Ekaterini Kotsopoulou, Marion Espeli, Lars Nitschke, Robert N Barker, Paul A Lyons, Kenneth G C Smith
ABSTRACT
CD22, an inhibitory co-receptor of the BCR, has been identified as a potential candidate gene for the development of autoimmune haemolytic anaemia in mice. In this study, we have examined Cd22(tm1Msn) CD22-deficient mice and identified an increase in RBC turnover and stress erythropoiesis, which might be consistent with haemolysis. We then, however, eliminated CD22 deficiency as the cause of accelerated RBC turnover and established that enhanced RBC turnover occurs independently of B cells and anti-RBC autoanti-bodies. Accelerated RBC turnover in this particular strain of CD22-deficient mice is red cell intrinsic and appears to be the consequence of a defective allele of glucose phosphate isomerase, Gpi1(c). This form of Gpi1 was originally derived from wild mice and results in a substantial reduction in enzyme activity. We have identified the polymorphism that causes impaired catalytic activity in the Gpi1(c) allele, and biochemically confirmed an approximate 75% reduction of GPI1 activity in Cd22(-/-) RBCs. The Cd22(-/-).Gpi1(c) congenic mouse provides a novel animal model of GPI1-deficiency, which is one of the most common causes of chronic non-spherocytic haemolytic anaemia in humans.
Product Description
Supelco
Phosphoglucose Isomerase from baker′s yeast (S. cerevisiae), for use with Fructose Assay Kit FA-20
Sigma-Aldrich
Phosphoglucose Isomerase from Bacillus stearothermophilus, lyophilized powder, 300-1,000 units/mg protein
Sigma-Aldrich
Phosphoglucose Isomerase from rabbit muscle, Type XI, lyophilized powder, ≥200 units/mg protein
Sigma-Aldrich
Phosphoglucose Isomerase from baker′s yeast (S. cerevisiae), Type III, ammonium sulfate suspension, ≥400 units/mg protein (biuret)