Merck
  • Effects of yeast hydrolysate on hepatic lipid metabolism in high-fat-diet-induced obese mice: yeast hydrolysate suppresses body fat accumulation by attenuating fatty acid synthesis.
Effects of yeast hydrolysate on hepatic lipid metabolism in high-fat-diet-induced obese mice: yeast hydrolysate suppresses body fat accumulation by attenuating fatty acid synthesis.
Annals of nutrition & metabolism (2012-08-15)
Eun Young Jung, Yang-Hee Hong, Jae Hwan Kim, Yooheon Park, Song Hwan Bae, Un Jae Chang, Hyung Joo Suh
ABSTRACT
We observed whether the anti-obesity activity of yeast hydrolysate (YH) was due to the alteration of lipid-regulating enzyme activities. Male ICR mice were divided into four groups: a normal diet group (ND; 4.2% fat), a high-fat diet group (HF; 27.7% fat), an HF group treated orally with 0.5% or 1% YH in the drinking water (HF+YH0.5; 27.7% fat and HF+YH1; 27.7% fat). After 5 weeks, the YH groups (HF+YH0.5=3.92±0.17 g/100 g BW and HF+YH1=3.76±0.13 g/100 g BW) had significantly lower levels of epididymal fats compared to the HF group (4.91±0.29 g/100 g BW; p<0.05). YH supplementation produced a decrease in serum triglycerides and low-density lipoprotein cholesterol concentrations and body weight gain, and produced a dose-dependent significant increase in serum ghrelin compared with the HF group (p<0.05). Hepatic glucose-6-phosphate dehydrogenase (G6PD) activity was inhibited by YH supplementation compared with the HF group, and mice treated orally with 1% YH exhibited a significant decrease in hepatic malic enzyme (ME) activity compared to obese mice treated with the vehicle (HF=10.44±2.74 nmol/min/mg protein vs. HF+YH1=6.68±2.23 nmol/min/mg protein; p<0.05). YH supplementation suppressed body fat accumulation by attenuating fatty acid synthesis through the downregulation of hepatic G6PD and ME activities.
Sigma-Aldrich
Malic Dehydrogenase from bovine heart, ammonium sulfate suspension, 2000-4000 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Malic Dehydrogenase from porcine heart, ≥600 units/mg protein (biuret), ammonium sulfate suspension
Sigma-Aldrich
Malic Dehydrogenase from porcine heart, ≥400 units/mg protein (biuret), ammonium sulfate suspension
Sigma-Aldrich
Malic Dehydrogenase from porcine heart, buffered aqueous glycerol solution, 600-1000 units/mg protein (biuret)