MilliporeSigma
  • Oral ingestion of aloe vera phytosterols alters hepatic gene expression profiles and ameliorates obesity-associated metabolic disorders in zucker diabetic fatty rats.
Oral ingestion of aloe vera phytosterols alters hepatic gene expression profiles and ameliorates obesity-associated metabolic disorders in zucker diabetic fatty rats.
Journal of agricultural and food chemistry (2012-02-23)
Eriko Misawa, Miyuki Tanaka, Kouji Nomaguchi, Kazumi Nabeshima, Muneo Yamada, Tomohiro Toida, Keiji Iwatsuki
ABSTRACT
We investigated the effects of the oral administration of lophenol (Lo) and cycloartanol (Cy), two kinds of antidiabetic phytosterol isolated from Aloe vera , on glucose and lipid metabolism in Zucker diabetic fatty (ZDF) rats. We demonstrated that the administrations of Lo and Cy suppressed random and fasting glucose levels and reduced visceral fat weights significantly. It was also observed that treatments with Lo and Cy decreased serum and hepatic lipid concentrations (triglyceride, nonesterified fatty acid, and total cholesterol). Additionally, Lo and Cy treatments resulted in a tendency for reduction in serum monocyte chemotactic protein-1 (MCP-1) level and an elevation in serum adiponectin level. Furthermore, the expression levels of hepatic genes encoding gluconeogenic enzymes (G6 Pase, PEPCK), lipogenic enzymes (ACC, FAS), and SREBP-1 were decreased significantly by the administrations of aloe sterols. In contrast, Lo and Cy administration increased mRNA levels of glycolysis enzyme (GK) in the liver. It was also observed that the hepatic β-oxidation enzymes (ACO, CPT1) and PPARα expressions tended to increase in the livers of the Lo- and Cy-treated rats compared with those in ZDF-control rats. We therefore conclude that orally ingested aloe sterols altered the expressions of genes related to glucose and lipid metabolism, and ameliorated obesity-associated metabolic disorders in ZDF rats. These findings suggest that aloe sterols could be beneficial in preventing and improving metabolic disorders with obesity and diabetes in rats.