Merck
  • Analysis of neofructooligosaccharides production mediated by the extracellular β-fructofuranosidase from Xanthophyllomyces dendrorhous.
Analysis of neofructooligosaccharides production mediated by the extracellular β-fructofuranosidase from Xanthophyllomyces dendrorhous.
Bioresource technology (2012-02-03)
Dolores Linde, Barbara Rodríguez-Colinas, Marta Estévez, Ana Poveda, Francisco J Plou, María Fernández Lobato
ABSTRACT
The extracellular β-fructofuranosidase Xd-INV from the yeast Xanthophyllomyces dendrorhous mainly synthesizes the neo-fructooligosaccharides (neo-FOS) neokestose and neonystose. This enzyme is a glycoprotein with a content of 59-67% N-linked carbohydrates and an estimated molecular mass of 160-200 kDa. The extent level of glycosylation affects the thermal behaviour of the enzyme but not its hydrolase and transferase activities, which are optimal at 60-70 °C. The neo-FOS yield of this enzyme was increased from 40 to 168 g/L when the sucrose concentration increased from 420 to 600 g/L and when the reaction was carried out at 60 °C. The neo-FOS levels obtained (168 g/L) in this work are the largest reported for any microbial β-fructofuranosidase.
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, BioReagent, ≥95% (SDS-PAGE), for proteomics
Sigma-Aldrich
PNGase F from Elizabethkingia miricola, buffered aqueous solution
Sigma-Aldrich
Glycopeptidase A from almonds, buffered aqueous glycerol solution, ≥0.05 unit/mL
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, lyophilized powder, recombinant, expressed in E. coli
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, recombinant, expressed in E. coli, set of 100 units nanomolar unit
Sigma-Aldrich
PNGase F from Elizabethkingia meningoseptica, ready-to-use solution, recombinant, expressed in E. coli