Merck
  • Biocatalytic potential of an alkalophilic and thermophilic dextranase as a remedial measure for dextran removal during sugar manufacture.
Biocatalytic potential of an alkalophilic and thermophilic dextranase as a remedial measure for dextran removal during sugar manufacture.
Bioresource technology (2012-01-27)
Shweta Purushe, Divya Prakash, Neelu N Nawani, Prashant Dhakephalkar, Balasaheb Kapadnis
ABSTRACT
The present study is focused on dextranase from Streptomyces sp. NK458 with potential to remove dextran formed during sugar manufacture. The dextranase had molecular weight of 130 kDa and hydrolyzed 15-25 and 410 kDa dextran. Dextranase production was optimized using statistical designs and the enzyme was purified 1.8-fold with 55.5% recovery. It displayed maximum activity at pH 9.0 and 60°C and was stable over a wide range of pH from 5.0 to 10.0. The k(m) and V(max) values were 3.05 mM and 17.97 mmol/ml/h, respectively. Ten units of dextranase could reduce dextran content by 67% in 24h and 56% in 72 h from sugarcane juice of cane variety CoS 86032. The enzyme was stable up to 3 days at 30°C beyond which its activity decreased and dextran removal could be retained by supplementation of 5 U of dextranase. These properties make it a promising biocatalyst for sugar industry.
Sigma-Aldrich
Dextranase from Chaetomium erraticum
Sigma-Aldrich
Dextranase from Penicillium sp., lyophilized powder, 100-250 units/mg protein
Sigma-Aldrich
Dextranase from Penicillium sp., lyophilized powder, 400-800 units/mg protein
Sigma-Aldrich
Dextranase from Penicillium sp., lyophilized powder, 10-25 units/mg solid
Sigma-Aldrich
IEF mix 3.6-9.3, lyophilized powder