Merck
  • Participation of the nonreducing terminal beta-galactosyl residues of the neutral N-linked carbohydrate chains of porcine zona pellucida glycoproteins in sperm-egg binding.
Participation of the nonreducing terminal beta-galactosyl residues of the neutral N-linked carbohydrate chains of porcine zona pellucida glycoproteins in sperm-egg binding.
Molecular reproduction and development (2004-12-01)
N Yonezawa, S Amari, K Takahashi, K Ikeda, F L Imai, S Kanai, K Kikuchi, M Nakano
ABSTRACT
The zona pellucida (ZP) surrounding the mammalian oocyte is composed of three glycoprotein components (ZPA, ZPB, and ZPC). Mammalian sperm bind to carbohydrate chains of a ZP glycoprotein in the initial phase of fertilization. Sperm-ligand carbohydrate chains have been characterized in mouse, cow, and pig. In pigs, triantennary/tetraantennary neutral complex-type chains from ZPB/ZPC mixture possess stronger sperm-binding activity than those of biantennary chains (Kudo et al., 1998: Eur J Biochem 252:492-499). Most of these oligosaccharides have beta-galactosyl residues at the nonreducing ends. This study used two in vitro competition assays to investigate the participation of the nonreducing terminal beta-galactosyl residues of the ligand active chains in porcine sperm binding. The removal of the nonreducing terminal beta-galactosyl residues from either the ligand active carbohydrate chains or endo-beta-galactosidase-digested glycoproteins significantly reduced their inhibition of sperm-egg binding, indicating that the beta-galactosyl residues at the nonreducing ends are involved in porcine sperm-egg binding. A correlation between the sperm-binding activity and in vitro fertilization rate is also presented.