Merck
  • Carbohydrate recognition by porphyrin-based molecularly imprinted polymers.
Carbohydrate recognition by porphyrin-based molecularly imprinted polymers.
Organic letters (2005-03-12)
Jung-Deog Lee, Nathaniel T Greene, Gregory T Rushton, Ken D Shimizu, Jong-In Hong
ABSTRACT
[structure: see text] Porphyrin-based molecularly imprinted polymers (MIPs) were prepared for carbohydrate recognition. A urea-appended porphyrin functional monomer was utilized to provide complementary functionality and quality binding sites throughout the polymer. Each porphyrin-based polymer demonstrates high affinity and differential selectivity for closely related carbohydrates that correlate to the structure of the template used in the imprinting process.