Merck
Tissue effect on genetic control of transcript isoform variation.
PLoS genetics (2009-08-15)
Tony Kwan, Elin Grundberg, Vonda Koka, Bing Ge, Kevin C L Lam, Christel Dias, Andreas Kindmark, Hans Mallmin, Osten Ljunggren, Fernando Rivadeneira, Karol Estrada, Joyce B van Meurs, Andre Uitterlinden, Magnus Karlsson, Claes Ohlsson, Dan Mellström, Olle Nilsson, Tomi Pastinen, Jacek Majewski
ABSTRACT
Current genome-wide association studies (GWAS) are moving towards the use of large cohorts of primary cell lines to study a disease of interest and to assign biological relevance to the genetic signals identified. Here, we use a panel of human osteoblasts (HObs) to carry out a transcriptomic survey, similar to recent studies in lymphoblastoid cell lines (LCLs). The distinct nature of HObs and LCLs is reflected by the preferential grouping of cell type-specific genes within biologically and functionally relevant pathways unique to each tissue type. We performed cis-association analysis with SNP genotypes to identify genetic variations of transcript isoforms, and our analysis indicates that differential expression of transcript isoforms in HObs is also partly controlled by cis-regulatory genetic variants. These isoforms are regulated by genetic variants in both a tissue-specific and tissue-independent fashion, and these associations have been confirmed by RT-PCR validation. Our study suggests that multiple transcript isoforms are often present in both tissues and that genetic control may affect the relative expression of one isoform to another, rather than having an all-or-none effect. Examination of the top SNPs from a GWAS of bone mineral density show overlap with probeset associations observed in this study. The top hit corresponding to the FAM118A gene was tested for association studies in two additional clinical studies, revealing a novel transcript isoform variant. Our approach to examining transcriptome variation in multiple tissue types is useful for detecting the proportion of genetic variation common to different cell types and for the identification of cell-specific isoform variants that may be functionally relevant, an important follow-up step for GWAS.
Sigma-Aldrich
GenElute Plant Genomic DNA Miniprep Kit, sufficient for 10 purifications
Sigma-Aldrich
GenElute Plant Genomic DNA Miniprep Kit, sufficient for 350 purifications
Sigma-Aldrich
GenElute Mammalian Genomic DNA Miniprep Kits, sufficient for 10 purifications
Sigma-Aldrich
GenElute Mammalian Genomic DNA Miniprep Kits, sufficient for 350 purifications
Sigma-Aldrich
GenElute Plant Genomic DNA Miniprep Kit, sufficient for 70 purifications
Sigma-Aldrich
GenElute Mammalian Genomic DNA Miniprep Kits, sufficient for 70 purifications
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, γ-irradiated by SER-TAIN process, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, γ-irradiated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, Heat Inactivated, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, suitable for cell culture
SAFC
Fetal Bovine Serum, Australia origin, γ-irradiated, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Dialyzed by ultrafiltration against 0.15 M NaCl, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, suitable for, Australia origin, USDA approved, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, Heat Inactivated, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for insect cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, sterile-filtered, non-USA origin, suitable for cell culture