Merck
The Drosophila fragile X mental retardation gene regulates sleep need.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2009-02-21)
Daniel Bushey, Giulio Tononi, Chiara Cirelli
ABSTRACT
Sleep need is affected by developmental stage and neuronal plasticity, but the underlying mechanisms remain unclear. The fragile X mental retardation gene Fmr1, whose loss-of-function mutation causes the most common form of inherited mental retardation in humans, is involved in synaptogenesis and synaptic plasticity, and its expression depends on both developmental stage and waking experience. Fmr1 is highly conserved across species and Drosophila mutants carrying dFmr1 loss-of-function or gain-of-function mutations are well characterized: amorphs have overgrown dendritic trees with larger synaptic boutons, developmental defects in pruning, and enhanced neurotransmission, while hypermorphs show opposite defects, including dendritic and axonal underbranching and loss of synapse differentiation. We find here that dFmr1 amorphs are long sleepers and hypermorphs are short sleepers, while both show increased locomotor activity and shortened lifespan. Both amorphs and hypermorphs also show abnormal sleep homeostasis, with impaired waking performance and no sleep rebound after sleep deprivation. An impairment in the circadian regulation of sleep cannot account for the altered sleep phenotype of dFmr1 mutants, nor can an abnormal activation of glutamatergic metabotropic receptors. Moreover, overexpression of dFmr1 throughout the mushroom bodies is sufficient to reduce sleep. Finally, dFmr1 protein levels are modulated by both developmental stage and behavioral state, with increased expression immediately after eclosure and after prolonged wakefulness. Thus, dFmr1 expression dose-dependently affects both sleep and synapses, suggesting that changes in sleep time in dFmr1 mutants may derive from changes in synaptic physiology.
Sigma-Aldrich
TWEEN® 20, viscosity 250-450 mPa.s (25 °C)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
TWEEN® 20, viscous liquid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
TWEEN® 20, Low-peroxide; Low-carbonyls
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
TWEEN® 20, viscous liquid, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
TWEEN® 20, for molecular biology, viscous liquid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
TWEEN® 20, Low-peroxide; Low-carbonyls
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
TWEEN® 20, BioXtra, viscous liquid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid